Merck
  • Lipolysis-derived linoleic acid drives beige fat progenitor cell proliferation.
Lipolysis-derived linoleic acid drives beige fat progenitor cell proliferation.
Developmental cell (2022-12-07)
Ichitaro Abe, Yasuo Oguri, Anthony R P Verkerke, Lauar B Monteiro, Carly M Knuth, Christopher Auger, Yunping Qiu, Gregory P Westcott, Saverio Cinti, Kosaku Shinoda, Marc G Jeschke, Shingo Kajimura
ABSTRACT
De novo beige adipocyte biogenesis involves the proliferation of progenitor cells in white adipose tissue (WAT); however, what regulates this process remains unclear. Here, we report that in mouse models but also in human tissues, WAT lipolysis-derived linoleic acid triggers beige progenitor cell proliferation following cold acclimation, β3-adrenoceptor activation, and burn injury. A subset of adipocyte progenitors, as marked by cell surface markers PDGFRα or Sca1 and CD81, harbored cristae-rich mitochondria and actively imported linoleic acid via a fatty acid transporter CD36. Linoleic acid not only was oxidized as fuel in the mitochondria but also was utilized for the synthesis of arachidonic acid-derived signaling entities such as prostaglandin D2. Oral supplementation of linoleic acid was sufficient to stimulate beige progenitor cell proliferation, even under thermoneutral conditions, in a CD36-dependent manner. Together, this study provides mechanistic insights into how diverse pathophysiological stimuli, such as cold and burn injury, promote de novo beige fat biogenesis.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Bovine Serum Albumin solution, 10% in DPBS, low endotoxin, fatty acid free, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, ≥99%, solid
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
MK-886, A cell-permeable, orally active NSAID that blocks cellular Cox pathway PGE2 production by inhibiting COX-1 and mPGES-1, but not COX-2, activity, as well as suppresses cellular 5-LO pathway activation by inhibiting FLAP, rather than 5-LO, activity.
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Baicalein, A cell-permeable flavone that inhibits the activity of 12-lipoxygenase (IC₅₀= 120 nM) and reverse transcriptase.