Merck
  • Engineered Upconversion Nanoparticles for Resolving Protein Interactions inside Living Cells.
Engineered Upconversion Nanoparticles for Resolving Protein Interactions inside Living Cells.
Angewandte Chemie (International ed. in English) (2016-08-12)
Christoph Drees, Athira Naduviledathu Raj, Rainer Kurre, Karin B Busch, Markus Haase, Jacob Piehler
ABSTRACT
Upconversion nanoparticles (UCNPs) convert near-infrared into visible light at much lower excitation densities than those used in classic two-photon absorption microscopy. Here, we engineered <50 nm UCNPs for application as efficient lanthanide resonance energy transfer (LRET) donors inside living cells. By optimizing the dopant concentrations and the core-shell structure for higher excitation densities, we observed enhanced UCNP emission as well as strongly increased sensitized acceptor fluorescence. For the application of these UCNPs in complex biological environments, we developed a biocompatible surface coating functionalized with a nanobody recognizing green fluorescent protein (GFP). Thus, rapid and specific targeting to GFP-tagged fusion proteins in the mitochondrial outer membrane and detection of protein interactions by LRET in living cells was achieved.
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4-Yb,Er@NaYF4, PEG-NH2 modified core-shell, 980 excitation, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, Oil soluble core shell,808 excitation, green
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4:Yb,Er@NaYF4, Oil soluble core-shell, fluorescence λex 980, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4,Yb,Nd, Oil soluble core shell upconversion nanoparticle, 808 excited, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4:Yb,Tm@NaYF4, Oil soluble core shell, 980 excitation, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4,Yb,Tm@NaYF4,Yb,Nd, 808 excited, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4-Yb,Tm@NaYF4, 980 excitation, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4-Yb,Er@NaYF4, PEG-COOH modified core-shell, fluorescence λex 980 nm, green light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4,Yb,Er@NaYF4,Yb,Nd, 808 excited, green
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4,Yb,Nd, PEG-COOH modified core-shell, fluorescence λex 808 nm, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4-Yb,Er@NaYF4, fluorescence λex 980 nm, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4 Yb, Nd, PEG-NH2 modified core-shell, fluorescence λex 808 nm, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, PEG-NH2 modified core-shell, 808 excitation, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, PEG-COOH modified core-shell, 808 excitation, green light