Merck
  • Membraneless condensates by Rapsn phase separation as a platform for neuromuscular junction formation.
Membraneless condensates by Rapsn phase separation as a platform for neuromuscular junction formation.
Neuron (2021-05-26)
Guanglin Xing, Hongyang Jing, Zheng Yu, Peng Chen, Hongsheng Wang, Wen-Cheng Xiong, Lin Mei
ABSTRACT
Our daily life depends on muscle contraction, a process that is controlled by the neuromuscular junction (NMJ). However, the mechanisms of NMJ assembly remain unclear. Here we show that Rapsn, a protein critical for NMJ formation, undergoes liquid-liquid phase separation (LLPS) and condensates into liquid-like assemblies. Such assemblies can recruit acetylcholine receptors (AChRs), cytoskeletal proteins, and signaling proteins for postsynaptic differentiation. Rapsn LLPS requires multivalent binding of tetratricopeptide repeats (TPRs) and is increased by Musk signaling. The capacity of Rapsn to condensate and co-condensate with interaction proteins is compromised by mutations of congenital myasthenic syndromes (CMSs). NMJ formation is impaired in mutant mice carrying a CMS-associated, LLPS-deficient mutation. These results reveal a critical role of Rapsn LLPS in forming a synaptic semi-membraneless compartment for NMJ formation.
Sigma-Aldrich
Mix-n-Stain CF 555 Antibody Labeling Kit (50-100μg)
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Mix-n-Stain CF 633 Antibody Labeling Kit (50-100μg)
Thrombin Protease, Cytiva 27-0846-01
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution