Merck
  • Host Tau Genotype Specifically Designs and Regulates Tau Seeding and Spreading and Host Tau Transformation Following Intrahippocampal Injection of Identical Tau AD Inoculum.
Host Tau Genotype Specifically Designs and Regulates Tau Seeding and Spreading and Host Tau Transformation Following Intrahippocampal Injection of Identical Tau AD Inoculum.
International journal of molecular sciences (2022-01-22)
Pol Andrés-Benito, Margarita Carmona, Mónica Jordán, Joaquín Fernández-Irigoyen, Enrique Santamaría, José Antoni Del Rio, Isidro Ferrer
ABSTRACT
Several studies have demonstrated the different characteristics of tau seeding and spreading following intracerebral inoculation in murine models of tau-enriched fractions of brain homogenates from AD and other tauopathies. The present study is centered on the importance of host tau in tau seeding and the molecular changes associated with the transformation of host tau into abnormal tau. The brains of three adult murine genotypes expressing different forms of tau-WT (murine 4Rtau), hTau (homozygous transgenic mice knock-out for murine tau protein and heterozygous expressing human forms of 3Rtau and 4Rtau proteins), and mtWT (homozygous transgenic mice knock-out for murine tau protein)-were analyzed following unilateral hippocampal inoculation of sarkosyl-insoluble tau fractions from the same AD and control cases. The present study reveals that (a) host tau is mandatory for tau seeding and spreading following tau inoculation from sarkosyl-insoluble fractions obtained from AD brains; (b) tau seeding does not occur following intracerebral inoculation of sarkosyl-insoluble fractions from controls; (c) tau seeding and spreading are characterized by variable genotype-dependent tau phosphorylation and tau nitration, MAP2 phosphorylation, and variable activation of kinases that co-localize with abnormal tau deposits; (d) transformation of host tau into abnormal tau is an active process associated with the activation of specific kinases; (e) tau seeding is accompanied by modifications in tau splicing, resulting in the expression of new 3Rtau and 4Rtau isoforms, thus indicating that inoculated tau seeds have the capacity to model exon 10 splicing of the host mapt or MAPT with a genotype-dependent pattern; (e) selective regional and cellular vulnerabilities, and different molecular compositions of the deposits, are dependent on the host tau of mice injected with identical AD tau inocula.
Sigma-Aldrich
Anti-phospho-Src (Tyr416) Antibody, clone 9A6, clone 9A6, Upstate®, from mouse
Sigma-Aldrich
Anti-cleaved-Tau (Asp421) Antibody, clone C3, clone C3, Upstate®, from mouse
Sigma-Aldrich
Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11, culture supernatant, clone 8E6/C11, Upstate®
Sigma-Aldrich
Anti-Tau (4-repeat isoform RD4) Antibody, clone 1E1/A6, culture supernatant, clone 1E1/A6, Upstate®
Sigma-Aldrich
Anti-phospho-NFL (Ser473) Antibody, clone 4F8, clone 4F8, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid