Merck
  • Expansion of tumor-associated Treg cells upon disruption of a CTLA-4-dependent feedback loop.
Expansion of tumor-associated Treg cells upon disruption of a CTLA-4-dependent feedback loop.
Cell (2021-06-23)
Francesco Marangoni, Ademi Zhakyp, Michela Corsini, Shannon N Geels, Esteban Carrizosa, Martin Thelen, Vinidhra Mani, Jasper N Prüßmann, Ross D Warner, Aleksandra J Ozga, Mauro Di Pilato, Shivashankar Othy, Thorsten R Mempel
ABSTRACT
Foxp3+ T regulatory (Treg) cells promote immunological tumor tolerance, but how their immune-suppressive function is regulated in the tumor microenvironment (TME) remains unknown. Here, we used intravital microscopy to characterize the cellular interactions that provide tumor-infiltrating Treg cells with critical activation signals. We found that the polyclonal Treg cell repertoire is pre-enriched to recognize antigens presented by tumor-associated conventional dendritic cells (cDCs). Unstable cDC contacts sufficed to sustain Treg cell function, whereas T helper cells were activated during stable interactions. Contact instability resulted from CTLA-4-dependent downregulation of co-stimulatory B7-family proteins on cDCs, mediated by Treg cells themselves. CTLA-4-blockade triggered CD28-dependent Treg cell hyper-proliferation in the TME, and concomitant Treg cell inactivation was required to achieve tumor rejection. Therefore, Treg cells self-regulate through a CTLA-4- and CD28-dependent feedback loop that adjusts their population size to the amount of local co-stimulation. Its disruption through CTLA-4-blockade may off-set therapeutic benefits in cancer patients.
Sigma-Aldrich
Anti-CD152 (CTLA-4) Antibody (mouse), APC, clone UC10-4F10-11, clone UC10-4F10-11, 0.2 mg/mL, from hamster(Armenian)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
IgG from rat serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder