Merck
  • Mutated KLF4(K409Q) in meningioma binds STRs and activates FGF3 gene expression.
Mutated KLF4(K409Q) in meningioma binds STRs and activates FGF3 gene expression.
iScience (2022-08-24)
Alla V Tsytsykova, Graham Wiley, Chuang Li, Richard C Pelikan, Lori Garman, Francis A Acquah, Blaine H M Mooers, Erdyni N Tsitsikov, Ian F Dunn
ABSTRACT
Krüppel-like factor 4 (KLF4) is a transcription factor that has been proven necessary for both induction and maintenance of pluripotency and self-renewal. Whole-genome sequencing defined a unique mutation in KLF4 (KLF4K409Q) in human meningiomas. However, the molecular mechanism of this tumor-specific KLF4 mutation is unknown. Using genome-wide high-throughput and focused quantitative transcriptional approaches in human cell lines, primary meningeal cells, and meningioma tumor tissue, we found that a change in the evolutionarily conserved DNA-binding domain of KLF4 alters its DNA recognition preference, resulting in a shift in downstream transcriptional activity. In the KLF4K409Q-specific targets, the normally silent fibroblast growth factor 3 (FGF3) is activated. We demonstrated a neomorphic function of KLF4K409Q in stimulating FGF3 transcription through binding to its promoter and in using short tandem repeats (STRs) located within the locus as enhancers.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
KLF4-TAT human, recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE), ≥90% (HPLC), suitable for cell culture
Sigma-Aldrich
Anti-SP3, C-Terminal antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-KLF4 antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
Sigma-Aldrich
Anti-Sp1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-KLF4 antibody produced in rabbit, affinity isolated antibody