Merck
  • Dual inhibition of autophagy and PI3K/mTOR pathway as a potential therapeutic strategy against laryngeal squamous cell carcinoma.
Dual inhibition of autophagy and PI3K/mTOR pathway as a potential therapeutic strategy against laryngeal squamous cell carcinoma.
Translational cancer research (2022-06-17)
Hui-Ying Huang, Ke-Nan Li, Hui-Ching Lau, Chi-Yao Hsueh, Ning Cong, Ming Zhang
ABSTRACT
New and effective chemotherapy or targeted therapy strategies are needed against laryngeal squamous cell carcinoma (LSCC). We aimed to explore the antitumor effect of dual PI3K/mTOR inhibitor combined with autophagy suppression on LSCC and its underlying mechanism. Hep-2 and AMC-HN-8 cell lines were treated with the Akt inhibitor LY294002, mTOR inhibitor rapamycin, and dual inhibitor NVP-BEZ235 separately. The biological characteristics of in vitro proliferation, cell cycle, apoptosis, migration, invasion, and autophagy were analyzed, and the expression levels of PI3K/Akt/mTOR pathway-related proteins were also measured. The in vivo effects of NVP-BEZ235 combined with inhibition of autophagy using pharmacological inhibitor was further assessed. Compared with Akt or mTOR inhibitor, NVP-BEZ235 had the most significant biological effects on LSCC cells. When combined with various autophagy inhibitors, along with siRNA against ATG7, NVP-BEZ235 showed a synergic antitumor effect in LSCC through increasing cell apoptosis and death both in vitro and vivo. NVP-BEZ235 exerted potent antitumor effects on LSCC, especially when combined with the autophagy inhibitor both in vitro and vivo, providing convincing experimental data for new molecular targeted therapy for LSCC.
Sigma-Aldrich
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-LC3 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-Akt (Ser473) Antibody, clone 6F5, clone 6F5, from mouse
Sigma-Aldrich
Anti-phospho-4E-BP1 (pThr70) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-P70S6K/RPS6KB1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-p70 S6 Kinase β (pSer423) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution