- Primary culture of tracheal aspirate-derived human airway basal stem cells.
Primary culture of tracheal aspirate-derived human airway basal stem cells.
STAR protocols (2022-05-24)
Gaurang M Amonkar, Richard Wagner, Kamakshi Bankoti, Jessica E Shui, Xingbin Ai, Paul H Lerou
PMID35600918
ABSTRACT
Patient-specific airway basal stem cells (BSCs) can be derived from tracheal aspirate (TA) samples from intubated patients, thus providing an invaluable lung stem cell derivation method that bypasses the need for lung tissue. The primary culture of BSCs provides the ideal model to study the function and differentiation of the conducting lung epithelium. This protocol outlines the specific steps for isolation, culture maintenance, passaging, freezing, thawing, differentiation, and immunofluorescence characterization of human TA-derived airway BSCs. For complete details on the use and execution of this protocol, please refer to Lu et al. (2021).
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-RFX3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Anti-SCGB1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution