Merck
  • Activation of AMPK by metformin promotes renal cancer cell proliferation under glucose deprivation through its interaction with PKM2.
Activation of AMPK by metformin promotes renal cancer cell proliferation under glucose deprivation through its interaction with PKM2.
International journal of biological sciences (2019-02-13)
Meihan Liu, Zhuo Zhang, Hui Wang, Xiaoliang Chen, Chunxiang Jin
ABSTRACT
Metformin, a common therapeutics for type 2 diabetics, was recently demonstrated to possess antitumor activity in various cancer types. However, its therapy effect in renal cell carcinoma (RCC) still remains controversial. In this study, we found that metformin treatment in RCC cells lead to activation of AMPK, which suppressed the cell proliferation under normal condition, but enhanced cell proliferation under glucose deprivation (GD) condition. Depletion of AMPK by siRNA abolished the proliferation effect of MF under GD condition. Mechanistic investigations revealed that the effect of AMPK on cell proliferation under GD condition is dependent on its nuclear translocation. Moreover, the nuclear AMPK recruits PKM2 and β-Catenin to form a complex, which promotes the transcription of cell proliferation related genes, including CCND1 and c-Myc. Furthermore, depletion of PKM2 or β-Catenin abrogated the proliferative effects of metformin under GD condition. And inhibition of PKM2 also re-sensitized the A498 xenograft in response to metformin treatment. Together, our results suggested that combined of AMPK activation and PKM2 depletion or inhibition can achieve better therapeutic effect for RCC patients.
Sigma-Aldrich
Anti-KI-67 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP6, tissue culture supernatant
Millipore
Protein G Plus/Protein A-Agarose, A mixture of Protein G PLUS and Protein A covalently conjugated to agarose. Useful for purification of IgG from biological fluids.
Sigma-Aldrich
Anti-AMPK α antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AMPK α (pThr172) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid