Merck
The IGFBP3/TMEM219 pathway regulates beta cell homeostasis.
Nature communications (2022-02-05)
Francesca D'Addio, Anna Maestroni, Emma Assi, Moufida Ben Nasr, Giovanni Amabile, Vera Usuelli, Cristian Loretelli, Federico Bertuzzi, Barbara Antonioli, Francesco Cardarelli, Basset El Essawy, Anna Solini, Ivan C Gerling, Cristina Bianchi, Gabriella Becchi, Serena Mazzucchelli, Domenico Corradi, Gian Paolo Fadini, Diego Foschi, James F Markmann, Emanuela Orsi, Jan Škrha, Maria Gabriella Camboni, Reza Abdi, A M James Shapiro, Franco Folli, Johnny Ludvigsson, Stefano Del Prato, Gianvincenzo Zuccotti, Paolo Fiorina
ABSTRACT
Loss of pancreatic beta cells is a central feature of type 1 (T1D) and type 2 (T2D) diabetes, but a therapeutic strategy to preserve beta cell mass remains to be established. Here we show that the death receptor TMEM219 is expressed on pancreatic beta cells and that signaling through its ligand insulin-like growth factor binding protein 3 (IGFBP3) leads to beta cell loss and dysfunction. Increased peripheral IGFBP3 was observed in established and at-risk T1D/T2D patients and was confirmed in T1D/T2D preclinical models, suggesting that dysfunctional IGFBP3/TMEM219 signaling is associated with abnormalities in beta cells homeostasis. In vitro and in vivo short-term IGFBP3/TMEM219 inhibition and TMEM219 genetic ablation preserved beta cells and prevented/delayed diabetes onset, while long-term IGFBP3/TMEM219 blockade allowed for beta cell expansion. Interestingly, in several patients' cohorts restoration of appropriate IGFBP3 levels was associated with improved beta cell function. The IGFBP3/TMEM219 pathway is thus shown to be a physiological regulator of beta cell homeostasis and is also demonstrated to be disrupted in T1D/T2D. IGFBP3/TMEM219 targeting may therefore serve as a therapeutic option in diabetes.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Human IGFBP-3 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Millipore
MILLIPLEX® Human IGF-I, II Magnetic Bead Panel - Endocrine Assay, The Human IGF Panel, using the Luminex xMAP technology, enables the simultaneous analysis of one or both IGF-I and IGF-II protein biomarkers in human serum, plasma and cell/tissue culture samples.
Sigma-Aldrich
Insulin-like Growth Factor-I human, IGF-I, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Millipore
MILLIPLEX® Human IGF Binding Protein Magnetic Bead Panel - Metabolism Multiplex Assay, The Human IGF Binding Protein Panel, using the Luminex xMAP technology, enables the simultaneous analysis of 7 IGFBP protein biomarkers in human serum, plasma and cell culture samples.
Roche
Cell Death Detection ELISA, sufficient for ≤96 tests, suitable for ELISA, kit of 1 (9 components)
Sigma-Aldrich
Anti-TMEM219 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution