Merck
  • Transcriptomics-based drug repositioning pipeline identifies therapeutic candidates for COVID-19.
Transcriptomics-based drug repositioning pipeline identifies therapeutic candidates for COVID-19.
Research square (2021-04-07)
Brian L Le, Gaia Andreoletti, Tomiko Oskotsky, Albert Vallejo-Gracia, Romel Rosales, Katharine Yu, Idit Kosti, Kristoffer E Leon, Daniel G Bunis, Christine Li, G Renuka Kumar, Kris M White, Adolfo García-Sastre, Melanie Ott, Marina Sirota
ABSTRACT
The novel SARS-CoV-2 virus emerged in December 2019 and has few effective treatments. We applied a computational drug repositioning pipeline to SARS-CoV-2 differential gene expression signatures derived from publicly available data. We utilized three independent published studies to acquire or generate lists of differentially expressed genes between control and SARS-CoV-2-infected samples. Using a rank-based pattern matching strategy based on the Kolmogorov-Smirnov Statistic, the signatures were queried against drug profiles from Connectivity Map (CMap). We validated sixteen of our top predicted hits in live SARS-CoV-2 antiviral assays in either Calu-3 or 293T-ACE2 cells. Validation experiments in human cell lines showed that 11 of the 16 compounds tested to date (including clofazimine, haloperidol and others) had measurable antiviral activity against SARS-CoV-2. These initial results are encouraging as we continue to work towards a further analysis of these predicted drugs as potential therapeutics for the treatment of COVID-19.
Product Description
USP
Fluticasone propionate, United States Pharmacopeia (USP) Reference Standard
Bacampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Dicycloverine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Lansoprazole, United States Pharmacopeia (USP) Reference Standard
Metixene hydrochloride, European Pharmacopoeia (EP) Reference Standard