Merck
  • Enhanced cGAS-STING-dependent interferon signaling associated with mutations in ATAD3A.
Enhanced cGAS-STING-dependent interferon signaling associated with mutations in ATAD3A.
The Journal of experimental medicine (2021-08-14)
Alice Lepelley, Erika Della Mina, Erika Van Nieuwenhove, Lise Waumans, Sylvie Fraitag, Gillian I Rice, Ashish Dhir, Marie-Louise Frémond, Mathieu P Rodero, Luis Seabra, Edwin Carter, Christine Bodemer, Daniela Buhas, Bert Callewaert, Pascale de Lonlay, Lien De Somer, David A Dyment, Fran Faes, Lucy Grove, Simon Holden, Marie Hully, Manju A Kurian, Hugh J McMillan, Kristin Suetens, Henna Tyynismaa, Stéphanie Chhun, Timothy Wai, Carine Wouters, Brigitte Bader-Meunier, Yanick J Crow
ABSTRACT
Mitochondrial DNA (mtDNA) has been suggested to drive immune system activation, but the induction of interferon signaling by mtDNA has not been demonstrated in a Mendelian mitochondrial disease. We initially ascertained two patients, one with a purely neurological phenotype and one with features suggestive of systemic sclerosis in a syndromic context, and found them both to demonstrate enhanced interferon-stimulated gene (ISG) expression in blood. We determined each to harbor a previously described de novo dominant-negative heterozygous mutation in ATAD3A, encoding ATPase family AAA domain-containing protein 3A (ATAD3A). We identified five further patients with mutations in ATAD3A and recorded up-regulated ISG expression and interferon α protein in four of them. Knockdown of ATAD3A in THP-1 cells resulted in increased interferon signaling, mediated by cyclic GMP-AMP synthase (cGAS) and stimulator of interferon genes (STING). Enhanced interferon signaling was abrogated in THP-1 cells and patient fibroblasts depleted of mtDNA. Thus, mutations in the mitochondrial membrane protein ATAD3A define a novel type I interferonopathy.
Sigma-Aldrich
Q-VD-OPh hydrate, ≥95% (HPLC)
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
DIDS, Disodium Salt, Binds covalently and irreversibly to the outer surface of human erythrocyte membranes.