Merck
  • Rab6A as a Pan-Astrocytic Marker in Mouse and Human Brain, and Comparison with Other Glial Markers (GFAP, GS, Aldh1L1, SOX9).
Rab6A as a Pan-Astrocytic Marker in Mouse and Human Brain, and Comparison with Other Glial Markers (GFAP, GS, Aldh1L1, SOX9).
Cells (2021-01-21)
Linda Melzer, Thomas M Freiman, Amin Derouiche
ABSTRACT
Astrocytes contribute to many higher brain functions. A key mechanism in glia-to-neuron signalling is vesicular exocytosis; however, the identity of exocytosis organelles remains a matter of debate. Since vesicles derived from the trans-Golgi network (TGN) are not considered in this context, we studied the astrocyte TGN by immunocytochemistry applying anti-Rab6A. In mouse brain, Rab6A immunostaining is found to be unexpectedly massive, diffuse in all regions, and is detected preferentially and abundantly in the peripheral astrocyte processes, which is hardly evident without glial fibrillary acid protein (GFAP) co-staining. All cells positive for the astrocytic markers glutamine synthetase (GS), GFAP, aldehyde dehydrogenase 1 family member L1 (Aldh1L1), or SRY (sex determining region Y)-box 9 (SOX9) were Rab6A+. Rab6A is excluded from microglia, oligodendrocytes, and NG2 cells using cell type-specific markers. In human cortex, Rab6A labelling is very similar and associated with GFAP+ astrocytes. The mouse data also confirm the specific astrocytic labelling by Aldh1L1 or SOX9; the astrocyte-specific labelling by GS sometimes debated is replicated again. In mouse and human brain, individual astrocytes display high variability in Rab6A+ structures, suggesting dynamic regulation of the glial TGN. In summary, Rab6A expression is an additional, global descriptor of astrocyte identity. Rab6A might constitute an organelle system with a potential role of Rab6A in neuropathological and physiological processes.
Sigma-Aldrich
Anti-ALDH1L1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Monoclonal Anti-RAB6A antibody produced in mouse, clone 3G3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit