Merck
  • Roles of NANOGP8 in cancer metastasis and cancer stem cell invasion during development of castration-resistant prostate cancer.
Roles of NANOGP8 in cancer metastasis and cancer stem cell invasion during development of castration-resistant prostate cancer.
Annals of translational medicine (2021-02-09)
Yi Sui, Wei Zhang, Rujian Zhu, Lili Gao, Ting Cao, Chuhong Chen, Min Gong, Hongbo Zhu, Tao Tang, Bo Yu, Tao Yang
ABSTRACT
Prostate cancer (PCa) is one of the most common types of cancer and the emerging resistance to androgen deprivation therapy in PCa aggravates disease progression. In this study, we examined the potential pro-tumorigenic functions of NANOGP8 in prostate cancer development. Quantitative RT-PCR confirmed higher NANOGP8 expression in androgen independent tumors, as well as a recurrent prostate tumor in patient samples. We then established a novel two-way inducible NANOGP8-short hairpin RNA experimental system, in which the NANOGP8 expression was transiently induced by adding doxycycline in the diet of NOD/SCID mice. The knockdown of NANOGP8 inhibited implanted tumor growth and the progression of castration-resistant PCa. NANOGP8-deficient PCa cells lost their cancer stem cell and gene expression programs. To further investigate the functions of NANOGP8 in PCa stem cells, real-time cell tracking was used to monitor the cell division modes and differentiation patterns of NANOGP8+ cells. The expression level of NANOGP8 markedly influenced the cell division mode of NANOGP8+ PCa cells and was strongly correlated with their pluripotency, reflected by robust telomerase activity and longer telomere length. NANOGP8 expression was also associated with the metastatic capacity of PCa cells. Based on these findings, we propose that NANOGP8 could serve as an effective therapeutic target for the treatment of PCa.
Sigma-Aldrich
Anti-beta-Actin Antibody, clone RM112, clone RM112, from rabbit
Sigma-Aldrich
Anti-Integrin α2β1 Antibody, clone BHA2.1, azide free, clone BHA2.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-NANOG Antibody, clone 7F7.1, clone 7F7.1, from mouse
Sigma-Aldrich
Anti-Vimentin Antibody, clone 3CB2, clone 3CB2, from mouse
Sigma-Aldrich
Anti-Aldehyde dehydrogenase 1 (ALDH1A1) Antibody, from rabbit, purified by affinity chromatography