Merck
  • ChemBead Enabled High-Throughput Cross-Electrophile Coupling Reveals a New Complementary Ligand.
ChemBead Enabled High-Throughput Cross-Electrophile Coupling Reveals a New Complementary Ligand.
Chemistry (Weinheim an der Bergstrasse, Germany) (2021-07-08)
Ana L Aguirre, Nathan L Loud, Keywan A Johnson, Daniel J Weix, Ying Wang
ABSTRACT
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of aryl bromides with alkyl halides to HTE is enabled by AbbVie ChemBeads technology. By using this approach, we were able to quickly map out the reactivity space at a global level with a challenging array of 3×222 micromolar reactions. The observed hit rate (56 %) is competitive with other often-used HTE reactions and the results are scalable. A key to this level of success was the finding that bipyridine 6-carboxamidine (BpyCam), a ligand that had not previously been shown to be optimal in any reaction, is as general as the best-known ligands with complementary reactivity. Such "cryptic" catalysts may be common and modern HTE methods should facilitate the process of finding these catalysts.
Product Description
N-XantPhos Pd G4 ChemBeads
Sigma-Aldrich
GPhos Pd G3 ChemBeads
Sigma-Aldrich
(t-Bu)PhCPhos Pd G4 ChemBeads
Sigma-Aldrich
GPhos Pd G4 ChemBeads
Dichloro(p-cymene)ruthenium(II) dimer ChemBeads
Sigma-Aldrich
DPPF Pd G4 ChemBeads
Sigma-Aldrich
QPhos Pd G3 ChemBeads
Sigma-Aldrich
APhos Pd G4 ChemBeads
Sigma-Aldrich
PPh3 Pd G2 ChemBeads
Sigma-Aldrich
Cesium Fluoride ChemBeads, NSC 84270
Sigma-Aldrich
Tris(dibenzylideneacetone)dipalladium(0) ChemBeads
Sigma-Aldrich
DPPF ChemBeads
APhos Pd G3 ChemBeads
Sigma-Aldrich
Hexaammineruthenium(III) chloride ChemBeads
Sigma-Aldrich
Copper(I) Chloride ChemBeads, 15 wt% loading
Sigma-Aldrich
tBuXPhos ChemBeads
Sigma-Aldrich
GPhos ChemBeads
Sigma-Aldrich
Potassium tert-butoxide ChemBeads
Sigma-Aldrich
Sodium hydroxide ChemBeads
Sigma-Aldrich
DavePhos Pd G3 ChemBeads
Sigma-Aldrich
P(o-tol)3 Pd G2 ChemBeads
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide ChemBeads
Sigma-Aldrich
MorDalPhos Pd G4 ChemBeads
Sigma-Aldrich
RockPhos Pd G3 ChemBeads
Copper(I) iodide ChemBeads
Sigma-Aldrich
AdBrettPhos Pd G3 ChemBeads
Sigma-Aldrich
BpyCAM•HCl
Sigma-Aldrich
Sodium tert-butoxide ChemBeads
Sigma-Aldrich
RuPhos ChemBeads
SPhos Pd G2 ChemBeads