Merck
  • Rapid Killing and Biofilm Inhibition of Multidrug-Resistant Acinetobacter baumannii Strains and Other Microbes by Iodoindoles.
Rapid Killing and Biofilm Inhibition of Multidrug-Resistant Acinetobacter baumannii Strains and Other Microbes by Iodoindoles.
Biomolecules (2020-08-23)
Chaitany Jayprakash Raorane, Jin-Hyung Lee, Jintae Lee
ABSTRACT
Multi-drug resistant Acinetobacter baumannii is well-known for its rapid acclimatization in hospital environments. The ability of the bacterium to endure desiccation and starvation on dry surfaces for up to a month results in outbreaks of health care-associated infections. Previously, indole and its derivatives were shown to inhibit other persistent bacteria. We found that among 16 halogenated indoles, 5-iodoindole swiftly inhibited A. baumannii growth, constrained biofilm formation and motility, and killed the bacterium as effectively as commercial antibiotics such as ciprofloxacin, colistin, and gentamicin. 5-Iodoindole treatment was found to induce reactive oxygen species, resulting in loss of plasma membrane integrity and cell shrinkage. In addition, 5-iodoindole rapidly killed three Escherichia coli strains, Staphylococcus aureus, and the fungus Candida albicans, but did not inhibit the growth of Pseudomonas aeruginosa. This study indicates the mechanism responsible for the activities of 5-iodoindole warrants additional study to further characterize its bactericidal effects on antibiotic-resistant A. baumannii and other microbes.
Sigma-Aldrich
6-Bromoindole, 96%
Sigma-Aldrich
5-Bromoindole, 99%
Sigma-Aldrich
5-Chloroindole, 98%
Sigma-Aldrich
2,3-Dimethylindole, ≥97%
Sigma-Aldrich
7-Chloroindole, 97%
Sigma-Aldrich
7-Bromoindole, 96%
Sigma-Aldrich
7-Iodoindole, 97%
Sigma-Aldrich
4-Bromoindole, 96%
Sigma-Aldrich
5-Nitroisatin, 97%
Sigma-Aldrich
5-Iodoindole, 97%
Sigma-Aldrich
6-Chloroindole, 99%