Merck
  • Polyreactive Broadly Neutralizing B cells Are Selected to Provide Defense against Pandemic Threat Influenza Viruses.
Polyreactive Broadly Neutralizing B cells Are Selected to Provide Defense against Pandemic Threat Influenza Viruses.
Immunity (2020-10-24)
Jenna J Guthmiller, Linda Yu-Ling Lan, Monica L Fernández-Quintero, Julianna Han, Henry A Utset, Dalia J Bitar, Natalie J Hamel, Olivia Stovicek, Lei Li, Micah Tepora, Carole Henry, Karlynn E Neu, Haley L Dugan, Marta T Borowska, Yao-Qing Chen, Sean T H Liu, Christopher T Stamper, Nai-Ying Zheng, Min Huang, Anna-Karin E Palm, Adolfo García-Sastre, Raffael Nachbagauer, Peter Palese, Lynda Coughlan, Florian Krammer, Andrew B Ward, Klaus R Liedl, Patrick C Wilson
ABSTRACT
Polyreactivity is the ability of a single antibody to bind to multiple molecularly distinct antigens and is a common feature of antibodies induced upon pathogen exposure. However, little is known about the role of polyreactivity during anti-influenza virus antibody responses. By analyzing more than 500 monoclonal antibodies (mAbs) derived from B cells induced by numerous influenza virus vaccines and infections, we found mAbs targeting conserved neutralizing influenza virus hemagglutinin epitopes were polyreactive. Polyreactive mAbs were preferentially induced by novel viral exposures due to their broad viral binding breadth. Polyreactivity augmented mAb viral binding strength by increasing antibody flexibility, allowing for adaption to imperfectly conserved epitopes. Lastly, we found affinity-matured polyreactive B cells were typically derived from germline polyreactive B cells that were preferentially selected to participate in B cell responses over time. Together, our data reveal that polyreactivity is a beneficial feature of antibodies targeting conserved epitopes.
Product Description
Supelco
GC Stationary Phase, phase 1,2,3-tris(2-Cyanoethoxy)propane (TCEP), bottle of 50 g
Sigma-Aldrich
N-p-Tosyl-L-phenylalanine chloromethyl ketone, ≥97% (TLC), powder
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin-Free, Sterile-Liquid, Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin Free, is a large, multi-subunit, O2-carrying, metalloprotein that is used as a carrier protein in the production of antibodies.