Merck
Plant cell cultures as food-aspects of sustainability and safety.
Plant cell reports (2020-09-07)
Suvi T Häkkinen, Heli Nygren, Liisa Nohynek, Riitta Puupponen-Pimiä, Raija-Liisa Heiniö, Natalia Maiorova, Heiko Rischer, Anneli Ritala
ABSTRACT
Sustainability and safety aspects of plant cell cultures as food are presented. Applicability of dairy side streams as carbon source and use of natural growth enhancers in cultivation are shown. Biotechnologically produced cellular products are currently emerging to replace and add into the portfolio of agriculturally derived commodities. Plant cell cultures used for food could supplement current food production. However, still many aspects need to be resolved before this new food concept can enter the market. Issues related to sustainability and safety for human consumption are relevant for both consumers and regulators. In this study, two plant cell cultures, deriving from arctic bramble (Rubus arcticus) and birch (Betula pendula), were cultivated using lactose-rich dairy side streams as alternative carbon sources to replace sucrose. Biomasses were comparable to those of original plant cell culture media when up to 83% and 75% of the original sucrose was replaced by these side streams for arctic bramble and birch cell cultures, respectively. Furthermore, nutritional composition or sensory properties were not compromised. Synthetic plant growth regulators were replaced by natural components, such as coconut water and IAA for several subculture cycles. Finally, it was shown that only trace amounts of free growth regulators are present in the cells at the harvesting point and assessment by freshwater crustaceans assay indicated that toxicity of the cells was not exceeding that of traditionally consumed bilberry fruit.
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
Coconut water, suitable for plant cell culture
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
Supelco
Thidiazuron, PESTANAL®, analytical standard
Sigma-Aldrich
Thidiazuron, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline