Merck
  • Blood-Derived Plasma Protein Biomarkers for Alzheimer's Disease in Han Chinese.
Blood-Derived Plasma Protein Biomarkers for Alzheimer's Disease in Han Chinese.
Frontiers in aging neuroscience (2019-01-09)
Zaohuo Cheng, Jiajun Yin, Hongwei Yuan, Chunhui Jin, Fuquan Zhang, Zhiqiang Wang, Xiaowei Liu, Yue Wu, Tao Wang, Shifu Xiao
ABSTRACT
It is well known that Alzheimer's disease (AD) is one of the most common progressive neurodegenerative diseases; it begins gradually, and therefore no effective medicine is administered in the beginning. Thus, early diagnosis and prevention of AD are crucial. The present study focused on comparing the plasma protein changes between patients with AD and their healthy counterparts, aiming to explore a specific protein panel as a potential biomarker for AD patients in Han Chinese. Hence, we recruited and collected plasma samples from 98 AD patients and 101 elderly healthy controls from Wuxi and Shanghai Mental Health Centers. Using a Luminex assay, we investigated the expression levels of fifty plasma proteins in these samples. Thirty-two out of 50 proteins were found to be significantly different between AD patients and healthy controls (P < 0.05). Furthermore, an eight-protein panel that included brain-derived neurotrophic factor (BDNF), angiotensinogen (AGT), insulin-like growth factor binding protein 2 (IGFBP-2), osteopontin (OPN), cathepsin D, serum amyloid P component (SAP), complement C4, and prealbumin (transthyretin, TTR) showed the highest determinative score for AD and healthy controls (all P = 0.00). In conclusion, these findings suggest that a combination of eight plasma proteins can serve as a promising diagnostic biomarker for AD with high sensitivity and specificity in Han Chinese populations; the eight plasma proteins were proven important for AD diagnosis by further cross-validation studies within the AD cohort.
Product Description
Millipore
MILLIPLEX® Human Metabolic Hormone Magnetic Bead Panel - Metabolism Multiplex Assay, The Human Metabolic Hormone Panel, using the Luminex xMAP technology, enables the simultaneous analysis of 13 protein biomarkers in human serum, plasma and cell/tissue culture samples.
Millipore
MILLIPLEX® Human Neurodegenerative Disease Magnetic Bead Panel 2 - Neuroscience Multiplex Assay, The analytes available for this multiplex kit are: α1-Antitrypsin, Complement C4, CRP, MIP-4, PEDF, SAP.
Millipore
MILLIPLEX® Human IGF Binding Protein Magnetic Bead Panel - Metabolism Multiplex Assay, The Human IGF Binding Protein Panel, using the Luminex xMAP technology, enables the simultaneous analysis of 7 IGFBP protein biomarkers in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human Neurodegenerative Disease Magnetic Bead Panel 1 - Neuroscience Multiplex Assay, The analytes available for this multiplex kit are: α2-Macroglobulin, Apo Al, Apo CIII, Apo E, Complement C3 and Complement Factor H.
Millipore
MILLIPLEX® Human MMP Magnetic Bead Panel 2 - Immunology Multiplex Assay, Matrix Metalloproteinase Bead-Based Multiplex Assays using the Luminex technology enable the simultaneous analysis of multiple MMPs biomarkers in human serum, plasma and cell culture samples.