Merck
  • Synthesis and evaluation of aryl-substituted diarylpropionitriles, selective ligands for estrogen receptor beta, as positron-emission tomographic imaging agents.
Synthesis and evaluation of aryl-substituted diarylpropionitriles, selective ligands for estrogen receptor beta, as positron-emission tomographic imaging agents.
Bioorganic & medicinal chemistry (2009-04-11)
Byung Seok Moon, Kathryn E Carlson, John A Katzenellenbogen, Tae Hyun Choi, Dae Yoon Chi, Jung Young Kim, Gi Jeong Cheon, Hun Yeong Koh, Kyo Chul Lee, Gwangil An
ABSTRACT
We have investigated halogen-substituted non-steroidal estrogens with selective binding affinity for the estrogen receptor beta (ERbeta that might be used for imaging the levels of this ER-subtype in breast tumors by positron emission tomography (PET). Based on diarylpropionitrile (DPN, 1a), a compound previously reported that has a 72-fold binding selectivity for ERbeta, we developed a series of DPN analogs having methyl-, hydroxyl-, and halogen substituents, including fluoroethyl and fluoropropyl groups. In competitive radiometric binding assays with [(3)H]estradiol, all of these DPN analogs showed high ERbeta/ERalpha selectivity; while the selectivity varied, in some cases it reached nearly 300-fold (RBA: ERalpha, 0.023%; ERbeta, 6.25%). The absolute ERbeta binding affinities, however, were not sufficient to merit further consideration for developing these ligands as PET imaging agents.
Sigma-Aldrich
4-Methoxyphenylacetonitrile, 97%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%