Merck
  • Pharmacological Inhibition of Acid Sphingomyelinase Prevents Uptake of SARS-CoV-2 by Epithelial Cells.
Pharmacological Inhibition of Acid Sphingomyelinase Prevents Uptake of SARS-CoV-2 by Epithelial Cells.
Cell reports. Medicine (2020-11-10)
Alexander Carpinteiro, Michael J Edwards, Markus Hoffmann, Georg Kochs, Barbara Gripp, Sebastian Weigang, Constantin Adams, Elisa Carpinteiro, Anne Gulbins, Simone Keitsch, Carolin Sehl, Matthias Soddemann, Barbara Wilker, Markus Kamler, Thomas Bertsch, Karl S Lang, Sameer Patel, Gregory C Wilson, Silke Walter, Hartmut Hengel, Stefan Pöhlmann, Philipp A Lang, Johannes Kornhuber, Katrin Anne Becker, Syed A Ahmad, Klaus Fassbender, Erich Gulbins
ABSTRACT
The acid sphingomyelinase/ceramide system plays an important role in bacterial and viral infections. Here, we report that either pharmacological inhibition of acid sphingomyelinase with amitriptyline, imipramine, fluoxetine, sertraline, escitalopram, or maprotiline or genetic downregulation of the enzyme prevents infection of cultured cells or freshy isolated human nasal epithelial cells with severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) or vesicular stomatitis virus (VSV) pseudoviral particles (pp-VSV) presenting SARS-CoV-2 spike protein (pp-VSV-SARS-CoV-2 spike), a bona fide system mimicking SARS-CoV-2 infection. Infection activates acid sphingomyelinase and triggers a release of ceramide on the cell surface. Neutralization or consumption of surface ceramide reduces infection with pp-VSV-SARS-CoV-2 spike. Treating volunteers with a low dose of amitriptyline prevents infection of freshly isolated nasal epithelial cells with pp-VSV-SARS-CoV-2 spike. The data justify clinical studies investigating whether amitriptyline, a safe drug used clinically for almost 60 years, or other antidepressants that functionally block acid sphingomyelinase prevent SARS-CoV-2 infection.
Sigma-Aldrich
Desipramine hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
Maprotiline hydrochloride, >99% (HPLC), powder
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
Sertraline hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Escitalopram oxalate, ≥98% (HPLC), powder
Sigma-Aldrich
Imipramine hydrochloride, BioXtra, ≥99% (TLC)