Merck
  • Poly(NaSS) functionalization modulates the conformation of fibronectin and collagen type I to enhance osteoblastic cell attachment onto Ti6Al4V.
Poly(NaSS) functionalization modulates the conformation of fibronectin and collagen type I to enhance osteoblastic cell attachment onto Ti6Al4V.
Langmuir : the ACS journal of surfaces and colloids (2014-07-24)
Helena P Felgueiras, Sven D Sommerfeld, N Sanjeeva Murthy, Joachim Kohn, Véronique Migonney
ABSTRACT
Functionalization of surfaces with poly(sodium styrenesulfonate) (poly(NaSS)) has recently been found to enhance osteointegration of implantable materials. Radical polymerization of poly(NaSS) on titanium (Ti)-based substrates has been used to improve their long-term performance by preventing fibrosis and consequently implant loosening. However, the influence of the sulfonate groups on the early cell behavior and the associated molecular phenomena remains to be understood. In this work, we used quartz crystal microbalance with dissipation (QCM-D) to elucidate the role of poly(NaSS) in enhancing osteoblastic cell attachment. This was measured by following the cell attachment using the MC3T3-E1 cell line, on fetal bovine serum (FBS) preadsorbed surfaces and on substrates adsorbed with a series of relevant proteins, bovine serum albumin (BSA), fibronectin (Fn), and collagen type I (Col I). Comparison of the performance of poly(NaSS) with other clinically important substrates such as Ti alloy Ti6Al4V, gold, and poly(desamino-tyrosyl-tyrosine ethyl ester carbonate) (poly(DTEc)) indicates poly(NaSS) to be a superior substrate for MC3T3-E1 cells attachment. This attachment was found to be integrin mediated in the presence of Fn and Col I. Antibodies specific to the RGD peptide and the N- and C-terminal HB-binding domains reacted more intensively with Fn adsorbed on poly(NaSS). Fn adapts a conformation favorable to RGD mediated cell attachment when adsorbed onto poly(NaSS).
Sigma-Aldrich
Anti-Fibronectin Antibody, cell binding peptide, clone 784A2A6, purified antibody, clone 784A2A6, Chemicon®
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone HMα2, clone HMalpha2, Chemicon®, from hamster(Armenian)
Sigma-Aldrich
Anti-Fibronectin Antibody, CT, clone 868A11, clone 868A11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin beta1 Antibody, clone HM beta 1.1, clone HM beta 1.1, from hamster