Merck
  • Histone Acetyltransferase MOF Blocks Acquisition of Quiescence in Ground-State ESCs through Activating Fatty Acid Oxidation.
Histone Acetyltransferase MOF Blocks Acquisition of Quiescence in Ground-State ESCs through Activating Fatty Acid Oxidation.
Cell stem cell (2020-07-02)
Le Tran Phuc Khoa, Yao-Chang Tsan, Fengbiao Mao, Daniel M Kremer, Peter Sajjakulnukit, Li Zhang, Bo Zhou, Xin Tong, Natarajan V Bhanu, Chunaram Choudhary, Benjamin A Garcia, Lei Yin, Gary D Smith, Thomas L Saunders, Stephanie L Bielas, Costas A Lyssiotis, Yali Dou
ABSTRACT
Self-renewing embryonic stem cells (ESCs) respond to environmental cues by exiting pluripotency or entering a quiescent state. The molecular basis underlying this fate choice remains unclear. Here, we show that histone acetyltransferase MOF plays a critical role in this process through directly activating fatty acid oxidation (FAO) in the ground-state ESCs. We further show that the ground-state ESCs particularly rely on elevated FAO for oxidative phosphorylation (OXPHOS) and energy production. Mof deletion or FAO inhibition induces bona fide quiescent ground-state ESCs with an intact core pluripotency network and transcriptome signatures akin to the diapaused epiblasts in vivo. Mechanistically, MOF/FAO inhibition acts through reducing mitochondrial respiration (i.e., OXPHOS), which in turn triggers reversible pluripotent quiescence specifically in the ground-state ESCs. The inhibition of FAO/OXPHOS also induces quiescence in naive human ESCs. Our study suggests a general function of the MOF/FAO/OXPHOS axis in regulating cell fate determination in stem cells.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
O-Acetyl-L-carnitine hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%
Sigma-Aldrich
Pyronin Y, certified by the Biological Stain Commission, Dye content 50 %
Sigma-Aldrich
1-(2,3,4-Trimethoxybenzyl)piperazine dihydrochloride, 97%
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys16) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Leukemia Inhibitory Factor human, LIF, recombinant, expressed in E. coli, 10 μg/ml, buffered aqueous solution (pH 7.4), suitable for cell culture
Sigma-Aldrich
Coenzyme A sodium salt hydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin solution, 7.5% in DPBS, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
PD 0325901, ≥98% (HPLC)
Sigma-Aldrich
LookOut® Mycoplasma PCR Detection Kit, Optimized for use with JumpStart Taq DNA Polymerase, D9307.
Sigma-Aldrich
Gö 6983, ≥97%
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®