Merck
  • Caspase-8 association with the focal adhesion complex promotes tumor cell migration and metastasis.
Caspase-8 association with the focal adhesion complex promotes tumor cell migration and metastasis.
Cancer research (2009-04-23)
Simone Barbero, Ainhoa Mielgo, Vicente Torres, Tal Teitz, David J Shields, David Mikolon, Matthew Bogyo, Daniela Barilà, Jill M Lahti, David Schlaepfer, Dwayne G Stupack
ABSTRACT
Caspase-8 is a proapoptotic protease that suppresses neuroblastoma metastasis by inducing programmed cell death. Paradoxically, caspase-8 can also promote cell migration among nonapoptotic cells; here, we show that caspase-8 can promote metastasis when apoptosis is compromised. Migration is enhanced by caspase-8 recruitment to the cellular migration machinery following integrin ligation. Caspase-8 catalytic activity is not required for caspase-8-enhanced cell migration; rather, caspase-8 interacts with a multiprotein complex that can include focal adhesion kinase and calpain 2 (CPN2), enhancing cleavage of focal adhesion substrates and cell migration. Caspase-8 association with CPN2/calpastatin disrupts calpastatin-mediated inhibition of CPN2. In vivo, knockdown of either caspase-8 or CPN2 disrupts metastasis among apoptosis-resistant tumors. This unexpected molecular collaboration provides an explanation for the continued or elevated expression of caspase-8 observed in many tumors.
Sigma-Aldrich
Collagen Type I, rat tail
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Leupeptin, synthetic, ≥85% (HPLC)
Sigma-Aldrich
ALLM, Cell permeable inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 600 pM).