Merck
Vascular Disease and Thrombosis in SARS-CoV-2-Infected Rhesus Macaques.
Cell (2020-10-17)
Malika Aid, Kathleen Busman-Sahay, Samuel J Vidal, Zoltan Maliga, Stephen Bondoc, Carly Starke, Margaret Terry, Connor A Jacobson, Linda Wrijil, Sarah Ducat, Olga R Brook, Andrew D Miller, Maciel Porto, Kathryn L Pellegrini, Maria Pino, Timothy N Hoang, Abishek Chandrashekar, Shivani Patel, Kathryn Stephenson, Steven E Bosinger, Hanne Andersen, Mark G Lewis, Jonathan L Hecht, Peter K Sorger, Amanda J Martinot, Jacob D Estes, Dan H Barouch
ABSTRACT
The COVID-19 pandemic has led to extensive morbidity and mortality throughout the world. Clinical features that drive SARS-CoV-2 pathogenesis in humans include inflammation and thrombosis, but the mechanistic details underlying these processes remain to be determined. In this study, we demonstrate endothelial disruption and vascular thrombosis in histopathologic sections of lungs from both humans and rhesus macaques infected with SARS-CoV-2. To define key molecular pathways associated with SARS-CoV-2 pathogenesis in macaques, we performed transcriptomic analyses of bronchoalveolar lavage and peripheral blood and proteomic analyses of serum. We observed macrophage infiltrates in lung and upregulation of macrophage, complement, platelet activation, thrombosis, and proinflammatory markers, including C-reactive protein, MX1, IL-6, IL-1, IL-8, TNFα, and NF-κB. These results suggest a model in which critical interactions between inflammatory and thrombosis pathways lead to SARS-CoV-2-induced vascular disease. Our findings suggest potential therapeutic targets for COVID-19.
Sigma-Aldrich
Anti-MxA, clone M143 (CL143), clone CL143, from mouse
Sigma-Aldrich
Monoclonal Anti-Collagen, Type I antibody produced in mouse, clone COL-1, ascites fluid
Sigma-Aldrich
Anti-CR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-C3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-IL6, affinity isolated antibody