Merck
  • Nonsteroidal anti-inflammatory drugs and their analogues as inhibitors of aldo-keto reductase AKR1C3: new lead compounds for the development of anticancer agents.
Nonsteroidal anti-inflammatory drugs and their analogues as inhibitors of aldo-keto reductase AKR1C3: new lead compounds for the development of anticancer agents.
Bioorganic & medicinal chemistry letters (2005-09-27)
Stanislav Gobec, Petra Brozic, Tea Lanisnik Rizner
ABSTRACT
Nonsteroidal anti-inflammatory drugs (NSAIDs) like indomethacin, flufenamic acid, and related compounds have been recently identified as potent inhibitors of AKR1C3. We report that some other NSAIDs (diclofenac and naproxen) also inhibit AKR1C3, with the IC(50) values in the low micromolar range. In order to obtain more information about the structure-activity relationship and to identify new leads, a series of compounds designed on the basis of NSAIDs were synthesized and screened on AKR1C3. The most active compounds were 2-[(2,2-diphenylacetyl)amino]benzoic acid 4 (IC(50)=11microM) and 3-phenoxybenzoic acid 10 (IC(50)=0.68microM). These compounds represent promising starting points for the development of new anticancer agents.
Sigma-Aldrich
4-Methoxybenzophenone, 97%
Supelco
Ibuprofen
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
2-Benzoylbenzoic acid, 98%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
4-Methylbenzophenone, 99%
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Supelco
Naproxen, VETRANAL®, analytical standard