Merck
  • Optoelectronic nose based on an origami paper sensor for selective detection of pesticide aerosols.
Optoelectronic nose based on an origami paper sensor for selective detection of pesticide aerosols.
Scientific reports (2020-10-16)
Mohammad Mahdi Bordbar, Tien-Anh Nguyen, Anh Quang Tran, Hasan Bagheri
ABSTRACT
This study introduces an applicable colorimetric sensor array for the detection of pesticides in the vapor phase. The array consisted of six metal nanoparticles spotted on the piece of filter paper. 3D-origami pattern was used for the fabrication of a paper-based sensor to decrease the effect of the nanoparticles leaching after exposure to analytes. Exposure to pesticide aerosols caused changes in the color of the array due to the aggregation of nanoparticles. These changes provided selective responses to thion pesticides such as malathion, parathion, chlorpyrifos, and diazinon. The sensing assay could also differentiate between aliphatic and aromatic thions and discriminate amine-containing compounds from the other studied analytes. These finding results are clearly confirmed by both visual detection and multivariate statistical methods. The proposed sensor was successfully developed for the quantitative measurement of pesticide aerosols at a very low concentration. The limit of detection of this method determined for malathion, parathion, chlorpyrifos and diazinon were 58.0, 103.0, 81.0 and 117.0, respectively. Moreover, the array could be employed to simultaneously analyze four studied pesticides. The statistcal results confirmed that the method has high performance for concurrent detection of thions as a major air pollutant without the interference of other species.
Sigma-Aldrich
2-Hydroxymethyl-1,3-propanediol, 97%
Sigma-Aldrich
Dimethylphenylphosphine, 99%