Merck
The Human and Mouse Enteric Nervous System at Single-Cell Resolution.
Cell (2020-09-06)
Eugene Drokhlyansky, Christopher S Smillie, Nicholas Van Wittenberghe, Maria Ericsson, Gabriel K Griffin, Gokcen Eraslan, Danielle Dionne, Michael S Cuoco, Max N Goder-Reiser, Tatyana Sharova, Olena Kuksenko, Andrew J Aguirre, Genevieve M Boland, Daniel Graham, Orit Rozenblatt-Rosen, Ramnik J Xavier, Aviv Regev
ABSTRACT
The enteric nervous system (ENS) coordinates diverse functions in the intestine but has eluded comprehensive molecular characterization because of the rarity and diversity of cells. Here we develop two methods to profile the ENS of adult mice and humans at single-cell resolution: RAISIN RNA-seq for profiling intact nuclei with ribosome-bound mRNA and MIRACL-seq for label-free enrichment of rare cell types by droplet-based profiling. The 1,187,535 nuclei in our mouse atlas include 5,068 neurons from the ileum and colon, revealing extraordinary neuron diversity. We highlight circadian expression changes in enteric neurons, show that disease-related genes are dysregulated with aging, and identify differences between the ileum and proximal/distal colon. In humans, we profile 436,202 nuclei, recovering 1,445 neurons, and identify conserved and species-specific transcriptional programs and putative neuro-epithelial, neuro-stromal, and neuro-immune interactions. The human ENS expresses risk genes for neuropathic, inflammatory, and extra-intestinal diseases, suggesting neuronal contributions to disease.
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei EZ Prep, sufficient for 25 nuclei preparations (~1-10×107 cells/preparation)
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Magnesium acetate solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Anti-mCherry, from chicken
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
SB-505124 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial