Merck
  • Distinct effects on cAMP signaling of carbamazepine and its structural derivatives do not correlate with their clinical efficacy in epilepsy.
Distinct effects on cAMP signaling of carbamazepine and its structural derivatives do not correlate with their clinical efficacy in epilepsy.
European journal of pharmacology (2020-08-08)
Sara Krarup, Christoffer Mertz, Emil Jakobsen, Sandy E H Lindholm, Lars H Pinborg, Lasse K Bak
ABSTRACT
The antiepileptic sodium channel blocker, carbamazepine, has long been known to be able to attenuate cAMP signals. This could be of clinical importance since cAMP signaling has been shown to be involved in epileptogenesis and seizures. However, no information on the ability to affect cAMP signaling is available for the marketed structural derivatives, oxcarbazepine and eslicarbazepine acetate or their dominating metabolite, licarbazepine. Thus, we employed a HEK293 cell line stably expressing a cAMP biosensor to assess the effect of these two drugs on cAMP accumulation. We find that oxcarbazepine does not affect cAMP accumulation whereas eslicarbazepine acetate, surprisingly, is able to enhance cAMP accumulation. Since the transcription of ADCY8 (adenylyl cyclase isoform 8; AC8) has been found to be elevated in epileptic tissue from patients, we subsequently expressed AC8 in the HEK293 cells. In the AC8-expressing cells, oxcarbazepine was now able to attenuate whereas eslicarbazepine maintained its ability to increase cAMP accumulation. However, at all concentrations tested, licarbazepine demonstrated no effect on cAMP accumulation. Thus, we conclude that the effects exerted by carbamazepine and its derivatives on cAMP accumulation do not correlate with their clinical efficacy in epilepsy. However, this does not disqualify cAMP signaling per se as a potential disease-modifying drug target for epilepsy since more potent and selective inhibitors may be of therapeutic value.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
BIA 2-093, ≥98% (HPLC), solid
Sigma-Aldrich
Oxcarbazepine, ≥98% (HPLC), solid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Supelco
10,11-Dihydro-10-hydroxycarbamazepine, analytical standard