Merck
  • Enzyme-linked immunosorbent assay (ELISA) and blocking with bovine serum albumin (BSA)--not all BSAs are alike.
Enzyme-linked immunosorbent assay (ELISA) and blocking with bovine serum albumin (BSA)--not all BSAs are alike.
Journal of immunological methods (2012-06-27)
Yuhong Xiao, Stuart N Isaacs
ABSTRACT
The enzyme-linked immunosorbent assay (ELISA) is an extremely common and powerful laboratory technique for detecting proteins by antibodies. Researchers frequently use bovine serum albumin (BSA) as a blocking agent to prevent non-specific binding of antigens and antibodies to the microtiter well. While studying the interactions of the vaccinia virus complement control protein (VCP) with complement, we found non-specific binding of VCP to BSA and identify a BSA preparation that did not result in non-specific binding. This work draws attention to the fact that not all BSA preparations are alike. It also highlights the need to perform critical controls to ensure that ELISA reactants do not inappropriately bind to the blocking agent.
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, RIA and ELISA Grade
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin solution, 30%±2% in 0.85% sodium chloride, aseptically filled
Sigma-Aldrich
Bovine Serum Albumin solution, 30% in 0.85% sodium chloride, aseptically filled
Sigma-Aldrich
Bovine Serum Albumin, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin, Bovine Serum, Cohn Fraction, 30% Aqueous Solution
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Fatty Acid-Poor, Endotoxin-Free
Sigma-Aldrich
Albumin, Bovine Serum, 30% Aqueous Solution, Stabilizer-Free
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Modified Cohn, pH 5.2
Sigma-Aldrich
Albumin fraction V, (from bovine serum) for biochemistry
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Fatty Acid-Free
Sigma-Aldrich
Albumin, Bovine Serum, 30% Sterile-Filtered Aqueous Solution, Preservative-Free
Roche
Bovine Serum Albumin Fraction V, protease-free, from bovine serum