Merck
  • Spinal caspase-6 contributes to remifentanil-induced hyperalgesia via regulating CCL21/CXCR3 pathway in rats.
Spinal caspase-6 contributes to remifentanil-induced hyperalgesia via regulating CCL21/CXCR3 pathway in rats.
Neuroscience letters (2020-02-06)
Chunyan Wang, Qing Li, Zhen Jia, Haifang Zhang, Yize Li, Qi Zhao, Lin Su, Yang Yu, Rubin Xu
ABSTRACT
Neuroinflammation in the spinal cord is a pathological event in remifentanil-induced hyperalgesia (RIH), but its underlying molecular mechanisms remain unclear. Recent studies recapitulate the significance of the intracellular protease caspase-6 in the release of inflammatory mediators and synaptic plasticity in pathologic pain. Also, chemokine CCL21 is involved in microglia activation and nociceptive transduction. This study examined whether spinal caspase-6 is associated with RIH via CCL21 and its receptor CXCR3. The acute exposure to remifentanil (1 μg kg-1 min-1for 60 min) was used to establish RIH, verified by assessment of mechanical paw withdrawal threshold and thermal paw withdrawal latency. The caspase-6 inhibitor, a neutralizing antibody against CCL21 (anti-CCL21), a selective CXCR3 antagonist NBI-74330, recombinant caspase-6 and CCL21 were used for the investigation of pathogenesis as well as the prevention of hyperalgesia. The expression of caspase-6, CCL21 and CXCR3 was also evaluated by RT-qPCR and Western blot. This study discovered mechanical allodynia and thermal hyperalgesia along with the increase in the expression of spinal caspase-6 and CCL21/CXCR3 after remifentanil exposure. Central caspase-6 inhibition prevented behavioral RIH and spinal up-regulation of CCL21/CXCR3 level. Intrathecal anti-CCL21 injection reduced RIH and spinal expression of CXCR3. The delivery of recombinant caspase-6 facilitated acute nociceptive hypersensitivity and increased spinal CXCR3 release in naïve rats, reversing by co-application of anti-CCL21. Also, NBI-74330 attenuated RIH and exogenous CCL21-caused acute pain behaviors. This study highlighted that spinal caspase-6-mediated up-regulation of CCL21/CXCR3 is vital in the pathogenesis of RIH in rats.
Sigma-Aldrich
Caspase-6, Human, Recombinant, E. coli, Caspase-6, Human, Recombinant is expressed as a single polypeptide in E. coli. Fused at the N- and C- termini to a His•Tag sequence. Undergoes spontaneous autoprocessing to yield the active enzyme.