Merck
  • Quinoline Containing Side-chain Antimalarial Analogs: Recent Advances and Therapeutic Application.
Quinoline Containing Side-chain Antimalarial Analogs: Recent Advances and Therapeutic Application.
Current topics in medicinal chemistry (2020-01-28)
Mukesh C Joshi, Timothy J Egan
ABSTRACT
The side-chains of quinoline antimalarial agents are the major concern of focus to build novel and efficaciaous bioactive and clinical antimalarials. Bioative antimalarial analogs may play a critical role in pH trapping in the food vacuole of RBC's with the help of fragmented amino acid, thus lead to Ī²-hematin inhibition. Here, the authors tried to summarize a useful, comprehensive compilation of side-chain modified ACQs along with their synthesis, biophysical and therapeutic applications etc. of potent antiplasmodial agents and therefore, opening the door towards the potential clinical status.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ā‰„99.0% (HPLC)
Sigma-Aldrich
3,4,5,6-Tetrachlorophthalimide, 97%