Merck
  • Evaluation of in vitro models of stem cell-derived cardiomyocytes to screen for potential cardiotoxicity of chemicals.
Evaluation of in vitro models of stem cell-derived cardiomyocytes to screen for potential cardiotoxicity of chemicals.
Toxicology in vitro : an international journal published in association with BIBRA (2020-05-25)
Miaoying Shi, Nguyen T Tien, Laura de Haan, Jochem Louisse, Ivonne M C M Rietjens, Hans Bouwmeester
ABSTRACT
Cardiotoxicity is an important toxicological endpoint for chemical and drug safety assessment. The present study aims to evaluate two stemcell-based in vitro models for cardiotoxicity screening of chemicals. Eleven model compounds were used to evaluate responses of mouse embryonic stem cell-derived cardiomyocytes (mESC-CMs) using beating arrest as a readout and the analysis of electrophysiological parameters measured with a multi-electrode array (MEA) platform of human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs). Results revealed that the hiPSC-CM MEA assay responded to all compounds. The mESC-CM beating arrest assay was not responsive to potassium channel blockers and showed a lower sensitivity to sodium channel blockers and Na+/K+ ATPase inhibitors compared to the hiPSC-CM MEA assay. Calcium channel blockers and a β-adrenergic receptor agonist showed comparable potencies in both models. The in vitro response concentrations from hiPSC-CMs were highly concordant with human effective serum concentrations of potassium and sodium channel blockers. It is concluded that both in vitro models enable the cardiotoxicity screening with different applicability domains. The mESC-CM beating arrest assay may be used as a first step in a tiered approach while the hiPSC-CM MEA assay may be the best starting point for quantitative in vitro to in vivo extrapolations.
Sigma-Aldrich
Flecainide acetate salt
Sigma-Aldrich
Mexiletine hydrochloride, powder
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Amiodarone hydrochloride, ≥98%
Sigma-Aldrich
Moxifloxacin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Dofetilide, ≥98% (HPLC)
USP
Isoproterenol hydrochloride, United States Pharmacopeia (USP) Reference Standard
g-Strophanthin, phyproof® Reference Substance