Merck
  • Hypoxia Promotes Prostate Cancer Aggressiveness by Upregulating EMT-Activator Zeb1 and SK3 Channel Expression.
Hypoxia Promotes Prostate Cancer Aggressiveness by Upregulating EMT-Activator Zeb1 and SK3 Channel Expression.
International journal of molecular sciences (2020-07-10)
Fanny Bery, Sandy Figiel, Sana Kouba, Delphine Fontaine, Maxime Guéguinou, Marie Potier-Cartereau, Christophe Vandier, Roseline Guibon, Franck Bruyère, Gaëlle Fromont, Karine Mahéo
ABSTRACT
Hypoxia is a well-established feature of prostate cancer (PCa) and is associated with disease aggressiveness. The hypoxic microenvironment initiates multiple adaptive responses including epithelial-to-mesenchymal transition (EMT) and a remodeling of calcium homeostasis involved in cancer progression. In the present study, we identified a new hypoxia signaling pathway with a positive feedback loop between the EMT transcription factor Zeb1 and SK3, a Ca2+-activated K+ channel, which leads to amplifying store-operated Ca2+ entry. Zeb1 and SK3 channel were strongly upregulated by hypoxia both in vitro and ex vivo in organotypic cultures of human PCa. Taking into account the sensitivity of the SK3 channel to the membrane lipid composition, we identified lipids such as Ohmline (an alkyl ether lipid and SK3 inhibitor), linoleic acid (LA) and eicosapentaenoic acid (EPA) (fatty acids associated with indolent PCa), which were able to completely abrogate the hypoxia-induced changes in Zeb1 expression. Ultimately, better understanding of this new hypoxia-induced EMT pathway may allow to develop adjuvant therapeutic strategies, in order to control PCa aggressiveness and improve treatment outcomes.
Sigma-Aldrich
Methyl palmitate, ≥99% (capillary GC)
Sigma-Aldrich
Methyl linoleate, ≥98% (GC)
Sigma-Aldrich
CyPPA, ≥98% (HPLC), solid
Supelco
Methyl all-cis-5,8,11,14,17-eicosapentaenoate, analytical standard