Merck
  • GPR101 drives growth hormone hypersecretion and gigantism in mice via constitutive activation of Gs and Gq/11.
GPR101 drives growth hormone hypersecretion and gigantism in mice via constitutive activation of Gs and Gq/11.
Nature communications (2020-09-23)
Dayana Abboud, Adrian F Daly, Nadine Dupuis, Mohamed Ali Bahri, Asuka Inoue, Andy Chevigné, Fabien Ectors, Alain Plenevaux, Bernard Pirotte, Albert Beckers, Julien Hanson
ABSTRACT
Growth hormone (GH) is a key modulator of growth and GH over-secretion can lead to gigantism. One form is X-linked acrogigantism (X-LAG), in which infants develop GH-secreting pituitary tumors over-expressing the orphan G-protein coupled receptor, GPR101. The role of GPR101 in GH secretion remains obscure. We studied GPR101 signaling pathways and their effects in HEK293 and rat pituitary GH3 cell lines, human tumors and in transgenic mice with elevated somatotrope Gpr101 expression driven by the rat Ghrhr promoter (GhrhrGpr101). Here, we report that Gpr101 causes elevated GH/prolactin secretion in transgenic GhrhrGpr101 mice but without hyperplasia/tumorigenesis. We show that GPR101 constitutively activates not only Gs, but also Gq/11 and G12/13, which leads to GH secretion but not proliferation. These signatures of GPR101 signaling, notably PKC activation, are also present in human pituitary tumors with high GPR101 expression. These results underline a role for GPR101 in the regulation of somatotrope axis function.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
Reticulum Stain Kit
Sigma-Aldrich
Calphostin C from Cladosporium cladosporioides, ≥90% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Horse Serum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-Mouse-IgG - Atto 647N antibody produced in goat, contains 50% glycerol as stabilizer
Sigma-Aldrich
H-89 dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit