Merck
  • Labeling, detection and identification of newly synthesized proteomes with bioorthogonal non-canonical amino-acid tagging.
Labeling, detection and identification of newly synthesized proteomes with bioorthogonal non-canonical amino-acid tagging.
Nature protocols (2007-04-05)
Daniela C Dieterich, Jennifer J Lee, A James Link, Johannes Graumann, David A Tirrell, Erin M Schuman
ABSTRACT
A major aim of proteomics is the identification of proteins in a given proteome at a given metabolic state. This protocol describes the step-by-step labeling, purification and detection of newly synthesized proteins in mammalian cells using the non-canonical amino acid azidohomoalanine (AHA). In this method, metabolic labeling of newly synthesized proteins with AHA endows them with the unique chemical functionality of the azide group. In the subsequent click chemistry tagging reaction, azide-labeled proteins are covalently coupled to an alkyne-bearing affinity tag. After avidin-based affinity purification and on-resin trypsinization, the resulting peptide mixture is subjected to tandem mass spectrometry for identification. In combination with deuterated leucine-based metabolic colabeling, candidate proteins can be immediately validated. Bioorthogonal non-canonical amino-acid tagging can be combined with any subcellular fractionation, immunopurification or other proteomic method to identify specific subproteomes, thereby reducing sample complexity and enabling the identification of subtle changes in a proteome. This protocol can be completed in 5 days.
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-Fluor 545, for Copper-free Click Chemistry
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulfo-dibenzocyclooctyne-biotin conjugate, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-biotin conjugate, for Copper-free Click Chemistry