Merck
  • Solar-driven photocatalytic treatment as sustainable strategy to remove pesticide residues from leaching water.
Solar-driven photocatalytic treatment as sustainable strategy to remove pesticide residues from leaching water.
Environmental science and pollution research international (2019-12-29)
Marina Aliste, Gabriel Pérez-Lucas, Nuria Vela, Isabel Garrido, José Fenoll, Simón Navarro
ABSTRACT
We have demonstrated the potential leaching of eight compounds, one insecticide (flonicamid) and seven fungicides (myclobutanil, penconazole, boscalid, difenoconazole, trifloxystrobin, pyraclostrobin and fenpyroximate) trough a typical Mediterranean soil (Calcaric regosol). The concentrations found in leaching water were in all cases above the limit set by the EU in groundwater (0.1 μg L-1). For this, the efficiency of different homogeneous (photo-Fenton and photo-Fenton-like) and heterogeneous (ZnO and TiO2) photocatalytic systems was tested in deionized water to choose the most appropriate treatment to remove pesticide residues from leaching water. The efficiency was in the order: ZnO + S2O82- (pH 7) > TiO2+ S2O82- (pH 7) > ZnO (pH 7) > TiO2 (pH 7) > Fe3+ (pH 3) > Fe3+ (pH 5) > Fe2+ (pH 3) > Fe2+ (pH 5). Thus, in the subsequent experiment we focus on the efficacy of solar heterogeneous photocatalysis (ZnO/Na2S2O8 and TiO2/ Na2S2O8) on their removal from leaching water. A fast removal was observed for all pesticides at the end of the photoperiod, noticeably higher in the case of ZnO system, with the exception of flonicamid, a recalcitrant pesticide where the degradation rate only reached about 20% after 240 min of solar exposure. Although the mineralisation of the initial dissolved organic carbon was not complete due to the presence of interfering substances in the leaching water, the conversion rate under ZnO/Na2S2O8 treatment was about 1.3 times higher than using TiO2/Na2S2O8.
Product Description
Supelco
Myclobutanil, PESTANAL®, analytical standard
Supelco
Trifloxystrobin, PESTANAL®, analytical standard
Supelco
Fenpyroximate, PESTANAL®, analytical standard