Merck
  • A P53-Independent DNA Damage Response Suppresses Oncogenic Proliferation and Genome Instability.
A P53-Independent DNA Damage Response Suppresses Oncogenic Proliferation and Genome Instability.
Cell reports (2020-02-06)
Katerina D Fagan-Solis, Dennis A Simpson, Rashmi J Kumar, Luciano G Martelotto, Lisle E Mose, Naim U Rashid, Alice Y Ho, Simon N Powell, Y Hannah Wen, Joel S Parker, Jorge S Reis-Filho, John H J Petrini, Gaorav P Gupta
ABSTRACT
The Mre11-Rad50-Nbs1 complex is a DNA double-strand break sensor that mediates a tumor-suppressive DNA damage response (DDR) in cells undergoing oncogenic stress, yet the mechanisms underlying this effect are poorly understood. Using a genetically inducible primary mammary epithelial cell model, we demonstrate that Mre11 suppresses proliferation and DNA damage induced by diverse oncogenic drivers through a p53-independent mechanism. Breast tumorigenesis models engineered to express a hypomorphic Mre11 allele exhibit increased levels of oncogene-induced DNA damage, R-loop accumulation, and chromosomal instability with a characteristic copy number loss phenotype. Mre11 complex dysfunction is identified in a subset of human triple-negative breast cancers and is associated with increased sensitivity to DNA-damaging therapy and inhibitors of ataxia telangiectasia and Rad3 related (ATR) and poly (ADP-ribose) polymerase (PARP). Thus, deficiencies in the Mre11-dependent DDR drive proliferation and genome instability patterns in p53-deficient breast cancers and represent an opportunity for therapeutic exploitation.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
VE-821, ≥98% (HPLC)
Sigma-Aldrich
Insulin, Zinc, Human, Recombinant, P. pastoris
Sigma-Aldrich
GenomePlex® Single Cell Whole Genome Amplification Kit, Amplify genome of a single cell
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Camptothecin, phyproof® Reference Substance