Merck
  • Deep Oxidative Desulfurization of Fuels in the Presence of Brönsted Acidic Polyoxometalate-Based Ionic Liquids.
Deep Oxidative Desulfurization of Fuels in the Presence of Brönsted Acidic Polyoxometalate-Based Ionic Liquids.
Molecules (Basel, Switzerland) (2020-01-30)
Argam Akopyan, Ekaterina Eseva, Polina Polikarpova, Anastasia Kedalo, Anna Vutolkina, Aleksandr Glotov
ABSTRACT
Polyoxometalate-based ionic liquid hybrid materials with a pyridinium cation, containing Brönsted acid sites, were synthesized and used as catalysts for the oxidation of model and real diesel fuels. Keggin-type polyoxometalates with the formulae [PMo12O40]3-, [PVMo11O40]4-, [PV2Mo10O40]4-, [PW12O40]3- were used as anions. It was shown that increasing the acid site strength leads to an increase of dibenzothiophene conversion to the corresponding sulfone. The best results were obtained in the presence of a catalyst, containing a nicotinic acid derivative as cation and phosphomolybdate as anion. The main factors affecting the process consisting of catalyst dosage, temperature, reaction time, oxidant dosage were investigated in detail. Under optimal conditions full oxidation of dibenzothiophene and more than a 90% desulfurization degree of real diesel fuel (initial sulfur content of 2050 ppm) were obtained (the oxidation conditions: NK-1 catalyst, molar ratio H2O2:S 10:1, molar ratio S:Mo 8:1, 1 mL MeCN, 70 °C, 1 h). The synthesized catalysts could be used five times with a slight decrease in activity.
Sigma-Aldrich
Dibenzothiophene, 98%
Sigma-Aldrich
Thianaphthene, 98%
Sigma-Aldrich
4,6-Dimethyldibenzothiophene, 97%
Sigma-Aldrich
Dibenzyl sulfide, ≥95.0% (HPLC)
Sigma-Aldrich
4-Methyldibenzothiophene, 96%