Merck
  • Context Specificity in Causal Signaling Networks Revealed by Phosphoprotein Profiling.
Context Specificity in Causal Signaling Networks Revealed by Phosphoprotein Profiling.
Cell systems (2016-12-27)
Steven M Hill, Nicole K Nesser, Katie Johnson-Camacho, Mara Jeffress, Aimee Johnson, Chris Boniface, Simon E F Spencer, Yiling Lu, Laura M Heiser, Yancey Lawrence, Nupur T Pande, James E Korkola, Joe W Gray, Gordon B Mills, Sach Mukherjee, Paul T Spellman
ABSTRACT
Signaling networks downstream of receptor tyrosine kinases are among the most extensively studied biological networks, but new approaches are needed to elucidate causal relationships between network components and understand how such relationships are influenced by biological context and disease. Here, we investigate the context specificity of signaling networks within a causal conceptual framework using reverse-phase protein array time-course assays and network analysis approaches. We focus on a well-defined set of signaling proteins profiled under inhibition with five kinase inhibitors in 32 contexts: four breast cancer cell lines (MCF7, UACC812, BT20, and BT549) under eight stimulus conditions. The data, spanning multiple pathways and comprising ∼70,000 phosphoprotein and ∼260,000 protein measurements, provide a wealth of testable, context-specific hypotheses, several of which we experimentally validate. Furthermore, the data provide a unique resource for computational methods development, permitting empirical assessment of causal network learning in a complex, mammalian setting.
Sigma-Aldrich
Anti-IRS1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-ERRFI1 antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Rad50 Antibody, clone 13B3/2C6, clone 13B3/2C6, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-erbB-2/HER-2 (Tyr1248) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Tau Antibody, clone 5E2, clone 5E2, Upstate®, from mouse
Sigma-Aldrich
Anti-MGMT Antibody, clone MT3.1, clone MT3.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-PKCδ (Ser664) Antibody, Upstate®, from rabbit