Merck
Beta-sitosterol sensitizes MDA-MB-231 cells to TRAIL-induced apoptosis.
Acta pharmacologica Sinica (2008-02-27)
Cheol Park, Dong-oh Moon, Chung-ho Ryu, Byung tae Choi, Won ho Lee, Gi-young Kim, Yung hyun Choi
ABSTRACT
To investigate whether subtoxic concentration of beta-sitosterol (SITO) combined with TNF-related apoptosis-inducing ligand (TRAIL) induces apoptosis in TRAIL-resistant MDA-MB-231 breast cancer cells. Cell viability and growth were assessed by 3-(4,5-dimethyl-2-thiazolyl)-2,5-diphnyl-2H-tetrazolim bromide assays, chromatin condensation, release of lactate dehydrogenase (LDH), and Annexin V+ cells. The apoptosis-related proteins were detected by Western blotting. Treatment with TRAIL in combination with subtoxic concentrations of SITO sensitized MDA-MB-231 breast cancer cells to TRAIL-mediated apoptosis. The synergistic treatment induced chromatin condensation, DNA fragmentation, the release of LDH, and Annexin V+ cells. The indicators of apoptosis are correlated to the induction of caspase activities, which results in the cleavage of poly(ADP-ribose)polymerase. Both the cytotoxic effects and apoptotic characteristics induced by the synergistic treatment were significantly inhibited by a pan-caspase inhibitor z-VAD-fmk, demonstrating the important role of caspases. These results indicate that caspases are crucial regulators of apoptosis induced by the combined treatment of SITO and TRAIL in MDA-MB-231 cells. The synergistic treatment of SITO and TRAIL induces apoptosis, which can serve as a potential preventive and therapeutic agent.
Sigma-Aldrich
Z-VAD-FMK, solid
Pricing and availability is not currently available.
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Pricing and availability is not currently available.