Merck
  • Ni-Catalyzed C-H Arylation of Oxazoles and Benzoxazoles Using Pharmaceutically Relevant Aryl Chlorides and Bromides.
Ni-Catalyzed C-H Arylation of Oxazoles and Benzoxazoles Using Pharmaceutically Relevant Aryl Chlorides and Bromides.
The Journal of organic chemistry (2019-09-25)
Helen Larson, Danielle Schultz, Dipannita Kalyani
ABSTRACT
This manuscript details the development of the nickel-catalyzed arylation of oxazoles and benzoxazoles with aryl halides. A series of aryl, heteroaryl, and druglike electrophiles relevant to pharmaceutical applications were surveyed. The desired arylated products were obtained in synthetically useful yields using electronically and structurally varied aryl halides. The use of microscale high-throughput experimentation was essential for both the rapid identification of optimal reaction parameters and the investigation of the aryl halide scope.
Sigma-Aldrich
Bis(1,5-cyclooctadiene)nickel(0)
Sigma-Aldrich
1,2-Bis(diphenylphosphino)benzene, 97%
Sigma-Aldrich
(Oxydi-2,1-phenylene)bis(diphenylphosphine), 98%
Sigma-Aldrich
2-Methyl-2-butanol, anhydrous, ≥99%
Sigma-Aldrich
5-Methylbenzoxazole, 97%
Sigma-Aldrich
1,2-Bis(dicyclohexylphosphino)ethane
Sigma-Aldrich
1,4-Bis(diphenylphosphino)butane, 98%