Merck
  • Genetic deletion of TNF receptor suppresses excitatory synaptic transmission via reducing AMPA receptor synaptic localization in cortical neurons.
Genetic deletion of TNF receptor suppresses excitatory synaptic transmission via reducing AMPA receptor synaptic localization in cortical neurons.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2011-10-11)
Ping He, Qiang Liu, Jie Wu, Yong Shen
ABSTRACT
The distribution of postsynaptic glutamate receptors has been shown to be regulated by proimmunocytokine tumor necrosis factor α (TNF-α) signaling. The role of TNF-α receptor subtypes in mediating glutamate receptor expression, trafficking, and function still remains unclear. Here, we report that TNF receptor subtypes (TNFR1 and TNFR2) differentially modulate α-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid receptor (AMPAR) clustering and function in cultured cortical neurons. We find that genetic deletion of TNFR1 decreases surface expression and synaptic localization of the AMPAR GluA1 subunit, reduces the frequency of miniature excitatory postsynaptic current (mEPSC), and reduces AMPA-induced maximal whole-cell current. In addition, these results are not observed in TNFR2-deleted neurons. The decreased AMPAR expression and function in TNFR1-deleted cells are not significantly restored by short (2 h) or long (24 h) term exposure to TNF-α. In TNFR2-deleted cells, TNF-α promotes AMPAR trafficking to the synapse and increases mEPSC frequency. In the present study, we find no significant change in the GluN1 subunit of NMDAR clusters, location, and mEPSC. This includes applying or withholding the TNF-α treatment in both TNFR1- and TNFR2-deleted neurons. Our results indicate that TNF receptor subtype 1 but not 2 plays a critical role in modulating AMPAR clustering, suggesting that targeting TNFR1 gene might be a novel approach to preventing neuronal AMPAR-mediated excitotoxicity.
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography