Merck
Nonporous Silica Nanoparticles for Nanomedicine Application.
Nano today (2013-09-03)
Li Tang, Jianjun Cheng
ABSTRACT
Nanomedicine, the use of nanotechnology for biomedical applications, has potential to change the landscape of the diagnosis and therapy of many diseases. In the past several decades, the advancement in nanotechnology and material science has resulted in a large number of organic and inorganic nanomedicine platforms. Silica nanoparticles (NPs), which exhibit many unique properties, offer a promising drug delivery platform to realize the potential of nanomedicine. Mesoporous silica NPs have been extensively reviewed previously. Here we review the current state of the development and application of nonporous silica NPs for drug delivery and molecular imaging.
Sigma-Aldrich
Silica, monodisperse, non-porous, 20 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 150 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 300 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 50 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 2.0 μm
Sigma-Aldrich
Silica, Monodisperse, non-porous, 40 µm
Sigma-Aldrich
Silica, monodisperse, non-porous, 400 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 200 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 5 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 750 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 10 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 1.0 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 500 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 70 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 3.0 μm
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 900 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 500 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 150 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 300 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 150 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 900 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 500 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 700 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 700 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 300 nm particle size, 5 % (w/w) in ethanol, dispersion