Merck
  • Excitonic Properties of Chemically Synthesized 2D Organic-Inorganic Hybrid Perovskite Nanosheets.
Excitonic Properties of Chemically Synthesized 2D Organic-Inorganic Hybrid Perovskite Nanosheets.
Advanced materials (Deerfield Beach, Fla.) (2018-03-27)
Qi Zhang, Leiqiang Chu, Feng Zhou, Wei Ji, Goki Eda
ABSTRACT
2D organic-inorganic hybrid perovskites (OIHPs) represent a unique class of materials with a natural quantum-well structure and quasi-2D electronic properties. Here, a versatile direct solution-based synthesis of mono- and few-layer OIHP nanosheets and a systematic study of their electronic structure as a function of the number of monolayers by photoluminescence and absorption spectroscopy are reported. The monolayers of various OIHPs are found to exhibit high electronic quality as evidenced by high quantum yield and negligible Stokes shift. It is shown that the ground exciton peak blueshifts by ≈40 meV when the layer thickness reduces from bulk to monolayer. It is also shown that the exciton binding energy remains effectively unchanged for (C6 H5 (CH2 )2 NH3 )2 PbI4 with the number of layers. Similar trends are observed for (C4 H9 NH3 )2 PbI4 in contrast to the previous report. Further, the photoluminescence lifetime is found to decrease with the number of monolayers, indicating the dominant role of surface trap states in nonradiative recombination of the electron-hole pairs.
Sigma-Aldrich
Dodecylamine, 98%
Sigma-Aldrich
N,N-Dimethylpropane-1,3-diammonium iodide
Sigma-Aldrich
n-Dodecylammonium iodide
Sigma-Aldrich
Butane-1,4-diammonium iodide
Sigma-Aldrich
Hexane-1,6-diammonium iodide
Sigma-Aldrich
Di-iso-Propylammonium iodide
Sigma-Aldrich
n-Hexylammonium iodide
Sigma-Aldrich
Hexadecylamine, 98%
Sigma-Aldrich
Hexylamine, 99%
Sigma-Aldrich
Di-n-Propylammonium iodide
Sigma-Aldrich
n-Octylammonium iodide, Greatcell Solar®
Sigma-Aldrich
2- Methyloxyethylammonium iodide