Merck
Pathway of Maternal Serotonin to the Human Embryo and Fetus.
Endocrinology (2018-01-31)
Harvey J Kliman, Sarah B Quaratella, Alessandra C Setaro, Erin C Siegman, Zarrin T Subha, Reshef Tal, Kristin M Milano, Theodore L Steck
ABSTRACT
Serotonin [5-hydroxytryptamine (5-HT)] is essential to intrauterine development, but its source is debated. We used immunocytochemistry to gauge 5-HT, its biosynthetic enzyme tryptophan hydroxylase 1 (TPH1); an importer (serotonin transporter, 5-HTT/SERT/SLC6A); other transporters [P-glycoprotein 1 (P-gp/ABCB1), OCT3/SLC22A3, and gap junction connexin-43]; and the 5-HT degradative enzyme monoamine oxidase A (MAOA) in sections of placentas. In humans, 5-HT was faintly stained only in first-trimester trophoblasts, whereas TPH1 was not seen at any stage. SERT was expressed in syncytiotrophoblasts and, more strongly, in cytotrophoblasts. MAOA was prominent in syncytiotrophoblasts, OCT3 and gap junctions were stained in cytotrophoblasts, and P-gp was present at the apical surfaces of both epithelia. 5-HT added to cultured placental explants accumulated in the trophoblast epithelium and reached the villus core vessels. Trophoblast uptake was blocked by the SERT inhibitor escitalopram. Inhibition of gap junctions with heptanol prevented the accumulation of 5-HT in cytotrophoblasts, whereas blocking OCT3 with decynium-22 and P-gp with mitotane led to its accumulation in cytotrophoblasts. Reducing 5-HT destruction by inhibiting MAOA with clorgyline increased the accumulation of 5-HT throughout the villus. In the mouse fetus, intravascular platelets stained prominently for 5-HT at day 13.5, whereas the placenta and yolk sac endoderm were both negative. TPH1 was not detected, but SERT was prominent in these mouse tissues. We conclude that serotonin is conveyed from the maternal blood stream through syncytiotrophoblasts, cytotrophoblasts and the villus core to the fetus through a physiological pathway that involves at least SERT, gap junctions, P-gp, OCT3, and MAOA.
Sigma-Aldrich
1,1′-Diethyl-2,2′-cyanine iodide, 97%
Sigma-Aldrich
Anti-Serotonin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Rabbit serum
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SLC22A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TPH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ABCB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
N-Methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine hydrochloride, ≥97% (GC)