Merck
  • Identification of the Nicotinamide Salvage Pathway as a New Toxification Route for Antimetabolites.
Identification of the Nicotinamide Salvage Pathway as a New Toxification Route for Antimetabolites.
Cell chemical biology (2018-02-27)
Daniela Buonvicino, Francesca Mazzola, Federica Zamporlini, Francesco Resta, Giuseppe Ranieri, Emidio Camaioni, Mirko Muzzi, Riccardo Zecchi, Giuseppe Pieraccini, Christian Dölle, Massimo Calamante, Gianluca Bartolucci, Mathias Ziegler, Barbara Stecca, Nadia Raffaelli, Alberto Chiarugi
ABSTRACT
Interest in the modulation of nicotinamide adenine dinucleotide (NAD) metabolome is gaining great momentum because of its therapeutic potential in different human disorders. Suppression of nicotinamide salvage by nicotinamide phosphoribosyl transferase (NAMPT) inhibitors, however, gave inconclusive results in neoplastic patients because several metabolic routes circumvent the enzymatic block converging directly on nicotinamide mononucleotide adenylyl transferases (NMNATs) for NAD synthesis. Unfortunately, NMNAT inhibitors have not been identified. Here, we report the identification of Vacor as a substrate metabolized by the consecutive action of NAMPT and NMNAT2 into the NAD analog Vacor adenine dinucleotide (VAD). This leads to inhibition of both enzymes, as well as NAD-dependent dehydrogenases, thereby causing unprecedented rapid NAD depletion, glycolytic block, energy failure, and necrotic death of NMNAT2-proficient cancer cells. Conversely, lack of NMNAT2 expression confers complete resistance to Vacor. Remarkably, Vacor prompts VAD formation and growth suppression in NMNAT2-positive neuroblastoma and melanoma xenografts. Our data show the first evidence of harnessing the entire nicotinamide salvage pathway for antimetabolic strategies.
Sigma-Aldrich
Inosine Monophosphate Dehydrogenase Type II human, recombinant, expressed in E. coli
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
9-Aminoacridine, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
GMX1778, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
β-Nicotinamide mononucleotide, ≥95% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
FK866 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder