
Zeatin

Synonym(s):
6-(4-Hydroxy-3-methylbut-2-enylamino)purine
Empirical Formula (Hill Notation):
C10H13N5O
CAS Number:
Molecular Weight:
219.24
Products
Brand
Color
Formula Weight
Physical Form
Product Line
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing