Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Xylenol orange tetrasodium salt
Synonym(s):
Xylenol orange tetrasodium salt, 3,3′-Bis[
N,N
-bis(carboxymethyl)aminomethyl]-
o
-cresolsulfonephthalein tetrasodium salt
Empirical Formula (Hill Notation):
C
31
H
28
N
2
Na
4
O
13
S
CAS Number:
3618-43-7
Molecular Weight:
760.58
EC Number:
222-805-8
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Special Grade
ACS reagent (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.08677
Xylenol orange tetrasodium salt,
metal indicator, ACS, Reag. Ph Eur
Expand
View Pricing
398187
Xylenol orange tetrasodium salt,
ACS reagent
Expand
View Pricing