
Xylazine

Synonym(s):
N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine, Xylazine
Empirical Formula (Hill Notation):
C12H16N2S
CAS Number:
Molecular Weight:
220.33
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing